siRNA Details
Gene Symbol: Esrp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107423.1 | NP_001100893.1 | 14 | 2256 | ||
XM_006255496.2 | XP_006255558.1 | 14 | 2126 | |||
XM_008772499.1 | XP_008770721.1 | 13 | 1821 | |||
GenBank mRNA | BC166804.1 | - | 14 | 2114 | 7938923 |
