siRNA Details
Gene Symbol: Esrp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107423.1 | NP_001100893.1 | 8, 9 | 1177 | ||
XM_006255496.2 | XP_006255558.1 | 8, 9 | 1047 | |||
XM_008772499.1 | XP_008770721.1 | 7, 8 | 742 | |||
GenBank mRNA | BC166804.1 | - | 8, 9 | 1035 | 7938923 |
