siRNA Details
Gene Symbol: Chst4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107427.1 | NP_001100897.1 | 2 | 757 | ||
XM_006255586.2 | XP_006255648.1 | 2 | 756 | |||
XM_006255587.2 | XP_006255649.1 | 2 | 839 |
