siRNA Details
Gene Symbol: Mon1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107433.1 | NP_001100903.1 | 1 | 125 | ||
XM_008772515.1 | XP_008770737.1 | 1 | 240 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Mon1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107433.1 | NP_001100903.1 | 1 | 125 | ||
XM_008772515.1 | XP_008770737.1 | 1 | 240 |