siRNA Details
Gene Symbol: Brd9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107453.1 | NP_001100923.1 | 6 | 721 | ||
XM_006227785.2 | XP_006227847.1 | 6 | 1006 | |||
XM_006227786.2 | XP_006227848.1 | 6 | 1006 | |||
XM_006227787.2 | XP_006227849.1 | 6 | 1006 |
