siRNA Details
Gene Symbol: Zfp541 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003748775.3 | XP_003748823.1 | 7 | 3001 | ||
XM_006228357.2 | XP_006228419.1 | 8 | 3066 | |||
XM_006228358.2 | XP_006228420.1 | 7 | 2738 | |||
XM_008759027.1 | XP_008757249.1 | 9 | 3176 | |||
XM_008759028.1 | XP_008757250.1 | 7 | 2785 | |||
XM_008759029.1 | XP_008757251.1 | 8 | 3111 |
