siRNA Details
Gene Symbol: Rgs17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107459.1 | NP_001100929.1 | 5, 6 | 699 | ||
XM_006227854.2 | XP_006227916.1 | 4, 5 | 640 | |||
XM_006227855.2 | XP_006227917.1 | 4, 5 | 661 |
