siRNA Details
Gene Symbol: Shprh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107470.1 | NP_001100940.1 | 11 | 2701 | ||
XM_006227639.2 | XP_006227701.1 | 11 | 2745 | |||
XM_008758609.1 | XP_008756831.1 | 11 | 2745 |
