siRNA Details
Gene Symbol: Mboat7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134978.1 | NP_001128450.1 | 685 | |||
GenBank mRNA | BC169089.1 | - | 685 | 5598892 |
Gene Symbol: LOC103689982 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008758837.1 | XP_008757059.1 | 5 | 685 |
