siRNA Details
Gene Symbol: Zfp110 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024775.1 | NP_001019946.1 | 8 | 1587 | ||
XM_006228111.2 | XP_006228173.1 | 7 | 2318 | |||
XM_006228112.2 | XP_006228174.1 | 8 | 2151 | |||
GenBank mRNA | BC097420.1 | - | 8 | 1587 | 7454819 |
