siRNA Details
Gene Symbol: Npas1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107479.1 | NP_001100949.1 | 8 | 1080 | ||
XM_006228413.2 | XP_006228475.1 | 9 | 1208 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Npas1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107479.1 | NP_001100949.1 | 8 | 1080 | ||
XM_006228413.2 | XP_006228475.1 | 9 | 1208 |