siRNA Details
Gene Symbol: Strn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107480.2 | NP_001100950.2 | 15, 16 | 2467 | ||
NM_001161809.1 | NP_001155281.1 | 15, 16 | 2446 | |||
GenBank mRNA | BC168948.1 | - | 14, 15 | 1653 | 7308304 |
