siRNA Details
Gene Symbol: Strn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107480.2 | NP_001100950.2 | 17 | 2645 | ||
NM_001161809.1 | NP_001155281.1 | 17 | 2624 | |||
GenBank mRNA | BC168948.1 | - | 16 | 1831 | 7308304 | |
FQ230133.1 | - | 1 | 16 |
