siRNA Details
Gene Symbol: Zfp112 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107487.1 | NP_001100957.1 | 4 | 2707 | ||
XM_006228422.2 | XP_006228484.1 | 5 | 2871 | |||
XM_008758926.1 | XP_008757148.1 | 4 | 2731 |
