siRNA Details
Gene Symbol: Cic | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107490.3 | 10 | 2108 | |||
XM_008758933.1 | XP_008757155.1 | 11 | 5619 | |||
XM_008758934.1 | XP_008757156.1 | 11 | 5883 | |||
XM_008758935.1 | XP_008757157.1 | 11 | 5619 | |||
XM_008758936.1 | XP_008757158.1 | 11 | 5619 | |||
XM_008758937.1 | XP_008757159.1 | 11 | 5619 | |||
XM_008758938.1 | XP_008757160.1 | 10 | 2301 | |||
XM_008758939.1 | XP_008757161.1 | 10 | 2302 | |||
XM_008758940.1 | XP_008757162.1 | 10 | 2302 |
