siRNA Details
Gene Symbol: Zfp383 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006228746.2 | XP_006228808.1 | 1698 | |||
XM_006228747.2 | XP_006228809.1 | 1630 | ||||
XM_006228748.2 | XP_006228810.1 | 1745 | ||||
XM_006228749.2 | XP_006228811.1 | 1583 | ||||
XM_006228750.2 | XP_006228812.1 | 1801 | ||||
XM_006228751.2 | XP_006228813.1 | 1520 |
Gene Symbol: LOC103690114 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006228705.2 | XP_006228767.1 | 4 | 1698 | ||
XM_006228706.2 | XP_006228768.1 | 5 | 1630 | |||
XM_006228707.2 | XP_006228769.1 | 5 | 1745 | |||
XM_006228708.2 | XP_006228770.1 | 5 | 1583 | |||
XM_006228709.2 | XP_006228771.1 | 5 | 1801 | |||
XM_006228710.2 | XP_006228772.1 | 5 | 1520 |
