siRNA Details
Gene Symbol: LOC103690114 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006228705.2 | XP_006228767.1 | 1 | 106 |
Gene Symbol: Zfp383 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006228746.2 | XP_006228808.1 | 106 |
