siRNA Details
Gene Symbol: Cd22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107503.1 | NP_001100973.1 | 1 | 296 | ||
XM_006228828.2 | XP_006228890.1 | 3 | 332 | |||
XM_008759152.1 | XP_008757374.1 | 2 | 166 | |||
XM_008759153.1 | XP_008757375.1 | 2 | 166 | |||
XM_008759154.1 | XP_008757376.1 | 2 | 166 |
