siRNA Details
Gene Symbol: RGD1308428 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271233.1 | NP_001258162.1 | 7 | 2048 | ||
XM_006228848.2 | XP_006228910.1 | 8 | 2272 | |||
XM_006228850.2 | XP_006228912.1 | 8 | 2131 | |||
XM_006228851.2 | XP_006228913.1 | 8 | 2862 | |||
XM_006228852.2 | XP_006228914.1 | 9 | 1714 | |||
XM_008759155.1 | XP_008757377.1 | 8 | 2130 | |||
XM_008759156.1 | XP_008757378.1 | 8 | 3972 | |||
XM_008759157.1 | XP_008757379.1 | 9 | 2199 | |||
XM_008759158.1 | XP_008757380.1 | 9 | 2210 | |||
XM_008759159.1 | XP_008757381.1 | 8 | 2135 | |||
GenBank mRNA | FQ221885.1 | - | 5 | 563 | ||
FQ223021.1 | - | 4 | 434 |
