siRNA Details
Gene Symbol: RGD1308428 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271233.1 | NP_001258162.1 | 9 | 2453 | ||
XM_006228848.2 | XP_006228910.1 | 10 | 2677 | |||
XM_006228850.2 | XP_006228912.1 | 10 | 2536 | |||
XM_006228851.2 | XP_006228913.1 | 10 | 3267 | |||
XM_006228852.2 | XP_006228914.1 | 11 | 2119 | |||
XM_008759155.1 | XP_008757377.1 | 10 | 2535 | |||
XM_008759156.1 | XP_008757378.1 | 10 | 4377 | |||
XM_008759157.1 | XP_008757379.1 | 11 | 2604 | |||
XM_008759158.1 | XP_008757380.1 | 11 | 2615 | |||
XM_008759159.1 | XP_008757381.1 | 10 | 2540 | |||
GenBank mRNA | FQ221885.1 | - | 7 | 968 | ||
FQ223021.1 | - | 6 | 839 |
