siRNA Details
Gene Symbol: Chst8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107504.2 | NP_001100974.1 | 4 | 1605 | ||
XM_006228853.2 | XP_006228915.1 | 4 | 2490 | |||
XM_006228854.2 | XP_006228916.1 | 4 | 1916 | |||
GenBank mRNA | BC161905.1 | - | 4 | 1605 | 8367390 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_175140, AB106878, AF308145, AK005217, AK051290, and others.
Gene Ontology Categories:
