siRNA Details
Gene Symbol: Nudt19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004258.1 | NP_001004258.1 | 2 | 722 | ||
XM_008759161.1 | XP_008757383.1 | 3 | 1649 | |||
GenBank mRNA | BC079088.1 | - | 2 | 722 | 7123358 | |
FQ215228.1 | - | 2 | 817 |
