siRNA Details
Gene Symbol: Ap2a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107511.2 | NP_001100981.1 | 7 | 777 | ||
XM_006229062.2 | XP_006229124.1 | 6 | 2621 | |||
XM_006229063.2 | XP_006229125.1 | 6 | 2621 | |||
XM_006229064.2 | XP_006229126.1 | 7 | 885 | |||
XM_006229065.2 | XP_006229127.1 | 7 | 885 |
