siRNA Details
Gene Symbol: Nipa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107519.1 | NP_001100989.1 | 3, 4 | 298 | ||
XM_008759393.1 | XP_008757615.1 | 3, 4 | 218 | |||
XM_008759394.1 | XP_008757616.1 | 3, 4 | 329 |
