siRNA Details
Gene Symbol: Herc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107520.1 | NP_001100990.1 | 21 | 3358 | ||
XM_006229300.2 | XP_006229362.1 | 22 | 3546 | |||
XM_006229301.2 | XP_006229363.1 | 22 | 3574 | |||
XM_006229302.2 | XP_006229364.1 | 22 | 3531 | |||
XM_006229304.2 | XP_006229366.1 | 22 | 3546 |
