siRNA Details
Gene Symbol: Chd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107523.1 | NP_001100993.1 | 25 | 3814 | ||
XM_006229369.2 | XP_006229431.1 | 24 | 6806 | |||
XM_006229370.2 | XP_006229432.1 | 24 | 6011 | |||
XM_006229371.2 | XP_006229433.1 | 25 | 5671 | |||
XM_006229372.2 | XP_006229434.1 | 23 | 6698 | |||
XM_008759519.1 | XP_008757741.1 | 24 | 6806 |
