siRNA Details
Gene Symbol: Hapln3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008559.1 | NP_001008559.1 | 2 | 413 | ||
XM_006229397.2 | XP_006229459.1 | 3 | 1271 | |||
GenBank mRNA | AB186131.1 | - | 2 | 413 |
