siRNA Details
Gene Symbol: Hapln3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008559.1 | NP_001008559.1 | 1 | 79 | ||
XM_006229397.2 | XP_006229459.1 | 2 | 937 | |||
GenBank mRNA | AB186131.1 | - | 1 | 79 | ||
FQ229812.1 | - | 2 | 180 |
