siRNA Details
Gene Symbol: Fsd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191627.1 | NP_001178556.1 | 12 | 2155 | ||
XM_006229440.2 | XP_006229502.1 | 13 | 2359 | |||
XM_008759537.1 | XP_008757759.1 | 13 | 3576 | |||
XM_008759538.1 | XP_008757760.1 | 14 | 2749 | |||
GenBank mRNA | FQ223649.1 | - | 13 | 2307 |
