siRNA Details
Gene Symbol: Usp35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008759807.1 | XP_008758029.1 | 7 | 1799 | ||
XM_238899.7 | XP_238899.4 | 6 | 1602 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Usp35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008759807.1 | XP_008758029.1 | 7 | 1799 | ||
XM_238899.7 | XP_238899.4 | 6 | 1602 |