siRNA Details
Gene Symbol: Fchsd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107539.1 | NP_001101009.1 | 7 | 641 | ||
XM_008759720.1 | XP_008757942.1 | 8 | 833 | |||
XM_008759721.1 | XP_008757943.1 | 8 | 833 | |||
XM_008759722.1 | XP_008757944.1 | 8 | 833 |
