siRNA Details
Gene Symbol: Fchsd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107539.1 | NP_001101009.1 | 16 | 1740 | ||
XM_008759720.1 | XP_008757942.1 | 18 | 2004 | |||
XM_008759721.1 | XP_008757943.1 | 18 | 2004 | |||
XM_008759722.1 | XP_008757944.1 | 17 | 1932 |
