siRNA Details
Gene Symbol: Numa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100691.1 | NP_001094161.1 | 17 | 5229 | ||
XM_008759723.1 | XP_008757945.1 | 18 | 5106 | |||
GenBank mRNA | BC098753.1 | - | 6 | 709 |
