siRNA Details
Gene Symbol: Rnf121 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107540.2 | NP_001101010.1 | 8 | 847 | ||
XM_006229878.2 | XP_006229940.1 | 8 | 1072 | |||
XM_006229879.2 | XP_006229941.1 | 9 | 1190 |
