siRNA Details
Gene Symbol: Mrvi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105210.1 | NP_001098680.1 | 17 | 2461 | ||
NM_001105211.1 | NP_001098681.1 | 16 | 2214 | |||
XM_006230047.2 | XP_006230109.1 | 17 | 2459 | |||
XM_006230048.2 | XP_006230110.1 | 18 | 2645 | |||
XM_008759727.1 | XP_008757949.1 | 17 | 2561 | |||
XM_008759728.1 | XP_008757950.1 | 17 | 2780 | |||
XM_008759729.1 | XP_008757951.1 | 18 | 2242 |
