siRNA Details
Gene Symbol: St5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107547.1 | NP_001101017.1 | 4 | 1054 | ||
XM_006229976.2 | XP_006230038.1 | 4 | 2374 | |||
XM_006229977.2 | XP_006230039.1 | 4 | 969 | |||
XM_006229979.2 | XP_006230041.1 | 3 | 1469 | |||
XM_008759737.1 | XP_008757959.1 | 2 | 2139 | |||
XM_008759738.1 | XP_008757960.1 | 4 | 2256 | |||
XM_008759739.1 | XP_008757961.1 | 5 | 2360 | |||
XM_008759740.1 | XP_008757962.1 | 2 | 2139 |
