siRNA Details
Gene Symbol: Pdzd9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024777.2 | NP_001019948.2 | 3, 4 | 358 | ||
XM_008759744.1 | XP_008757966.1 | 4, 5 | 2817 | |||
GenBank mRNA | AY387091.1 | - | 3, 4 | 358 |
