siRNA Details
Gene Symbol: Tnpo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100692.1 | NP_001094162.1 | 3 | 230 | ||
XM_006231839.2 | XP_006231901.1 | 4 | 821 | |||
XM_006231840.2 | XP_006231902.1 | 4 | 545 | |||
XM_008760668.1 | XP_008758890.1 | 4 | 833 |

More Information
Other Species Targeted

Gene Ontology Categories:
