siRNA Details
Gene Symbol: Rce1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100585.1 | NP_001094055.1 | 5 | 507 | ||
XM_006230788.2 | XP_006230850.1 | 5 | 537 | |||
XM_006230790.2 | XP_006230852.1 | 5 | 537 | |||
XM_006230791.2 | XP_006230853.1 | 5 | 537 | |||
XM_008760135.1 | XP_008758357.1 | 5 | 562 | |||
XM_008760136.1 | XP_008758358.1 | 5 | 447 | |||
GenBank mRNA | BC087629.1 | - | 5 | 494 | ||
BC158593.1 | - | 5 | 507 | 8369432 |
