siRNA Details
Gene Symbol: Ehbp1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025117.2 | NP_001020288.2 | 12 | 1898 | ||
NM_001129997.1 | NP_001123469.1 | 12 | 4670 | |||
NM_001270704.1 | NP_001257633.1 | 12 | 2393 | |||
NM_001270705.1 | NP_001257634.1 | 11 | 1862 | |||
XM_008760139.1 | XP_008758361.1 | 12 | 2428 | |||
GenBank mRNA | BC087115.1 | - | 11 | 1825 | ||
BC129126.1 | - | 12 | 1898 | |||
BC167745.1 | - | 12 | 2174 | 7310903 |
