siRNA Details
Gene Symbol: Taf6l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107575.1 | NP_001101045.1 | 12 | 1890 | ||
XM_006230979.2 | XP_006231041.1 | 12 | 2109 | |||
XM_006230981.2 | XP_006231043.1 | 12 | 2052 | |||
XM_006230982.2 | XP_006231044.1 | 13 | 1979 | |||
XM_006230983.2 | XP_006231045.1 | 12 | 1890 | |||
XM_008760224.1 | XP_008758446.1 | 12 | 2361 | |||
GenBank mRNA | BC168911.1 | - | 12 | 2005 | 7458553 |
Gene Symbol: Tmem179b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006231001.2 | XP_006231063.1 | 1 | 697 |
