siRNA Details
Gene Symbol: Dagla | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005886.1 | NP_001005886.1 | 14 | 1482 | ||
XM_008760229.1 | XP_008758451.1 | 13 | 4852 | |||
XM_008760230.1 | XP_008758452.1 | 14 | 4767 | |||
GenBank mRNA | AY275375.1 | - | 14 | 1482 |
