siRNA Details
Gene Symbol: Vwce | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271311.1 | NP_001258240.1 | 2188 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Vwce | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271311.1 | NP_001258240.1 | 2188 |