siRNA Details
Gene Symbol: Glyatl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126278.1 | NP_001119750.1 | 5 | 617 | ||
GenBank mRNA | BC160862.1 | - | 5 | 617 | 7323546 | |
FQ210014.1 | - | 771 | ||||
FQ219712.1 | - | 5 | 620 |
