siRNA Details
Gene Symbol: Fan1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191633.1 | NP_001178562.1 | 1 | 782 | ||
XM_006229339.2 | XP_006229401.1 | 2 | 1209 | |||
XM_008759539.1 | XP_008757761.1 | 1 | 1389 |
