siRNA Details
Gene Symbol: Plxna3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107581.2 | NP_001101051.2 | 9 | 2018 | ||
XM_006229577.2 | XP_006229639.1 | 9 | 2177 | |||
XM_006229578.2 | XP_006229640.1 | 9 | 2434 | |||
XM_008773598.1 | XP_008771820.1 | 8 | 3926 | |||
XM_008773599.1 | XP_008771821.1 | 8 | 3926 |
