siRNA Details
Gene Symbol: Zfyve27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199104.2 | NP_954535.2 | 9, 10 | 1204 | ||
XM_006231392.2 | XP_006231454.1 | 10, 11 | 1241 | |||
XM_008760422.1 | XP_008758644.1 | 9, 10 | 1749 | |||
XM_008760423.1 | XP_008758645.1 | 8, 9 | 1728 | |||
GenBank mRNA | BC061741.1 | - | 9, 10 | 1170 | 5599023 |
