siRNA Details
Gene Symbol: Kank1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037197.2 | NP_001032274.2 | 4 | 1419 | ||
XM_006231212.2 | XP_006231274.1 | 2 | 2362 | |||
XM_006231213.2 | XP_006231275.1 | 2 | 4043 | |||
XM_006231214.2 | XP_006231276.1 | 4 | 1536 | |||
XM_006231215.2 | XP_006231277.1 | 3 | 1447 | |||
XM_006231216.2 | XP_006231278.1 | 4 | 3970 | |||
XM_006231218.2 | XP_006231280.1 | 2 | 2362 | |||
XM_006231219.1 | XP_006231281.1 | 2 | 1360 | |||
XM_006231220.1 | XP_006231282.1 | 2 | 1283 | |||
XM_008760305.1 | XP_008758527.1 | 4 | 1553 |
