siRNA Details
Gene Symbol: Dmrt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107597.1 | NP_001101067.1 | 1 | 295 | ||
XM_008760306.1 | XP_008758528.1 | 1 | 299 | |||
XM_008760307.1 | XP_008758529.1 | 1 | 299 |
