siRNA Details
Gene Symbol: Sec31b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135713.1 | NP_001129185.1 | 12 | 1534 | ||
XM_006231484.2 | XP_006231546.1 | 12 | 1642 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Sec31b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135713.1 | NP_001129185.1 | 12 | 1534 | ||
XM_006231484.2 | XP_006231546.1 | 12 | 1642 |